Propen-1-ylsulfur pentafluoride

Supplier: APOLLO SCIENTIFIC
Warning

PC450556-1G PC450556-250MG
77415-560EA 544.04 USD
77415-560 77415-562
Propen-1-ylsulfur pentafluoride
Propen-1-ylsulfur pentafluoride
Formula: C₃H₅F₅S
Storage Temperature: Ambient
MDL Number: MFCD31561016
CAS Number: 690-04-0
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR