2,2-Difluoroethyl hexafluoroisopropyl carbonate

Supplier: APOLLO SCIENTIFIC
Warning

PC450294-1G PC450294-5G
77415-542EA 185.92 USD
77415-542 77415-544
2,2-Difluoroethyl hexafluoroisopropyl carbonate
2,2-Difluoroethyl hexafluoroisopropyl carbonate
Formula: C₆H₄F₈O₃
MW: 276.081 g/mol
Boiling Pt: 80…81 °C (150 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD28334289
CAS Number: 1980044-06-1
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR