1H,1H-Heptafluorobutyl methyl ether 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 1,1,1,2,2,3,3-Heptafluoro-4-methoxybutane

PC450249-1G PC450249-5G
77415-508EA 83.54 USD
77415-508 77415-510
1H,1H-Heptafluorobutyl methyl ether 97%
1H,1H-Heptafluorobutyl methyl ether
Formula: C₅H₅F₇O
MW: 214.081 g/mol
Boiling Pt: 68 °C
Density: 1.3945 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD16621315
CAS Number: 376-98-7
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR