2-(2,2-Difluoroethoxy)ethanol

Supplier: APOLLO SCIENTIFIC
Warning

PC450198-25G PC450198-5G PC450198-1G
77415-494EA 908.78 USD
77415-494 77415-496 77415-492
2-(2,2-Difluoroethoxy)ethanol
2-(2,2-Difluoroethoxy)ethanol
Formula: C₄H₈F₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD19442036
CAS Number: 853005-07-9
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR