5,6-Dichloro-2-(trifluoromethyl)-1H-benzimidazole

Supplier: APOLLO SCIENTIFIC
Danger

PC450181-1G PC450181-5G
77415-468EA 245.54 USD
77415-468 77415-470
5,6-Dichloro-2-(trifluoromethyl)-1H-benzimidazole
5,6-Dichloro-2-(trifluoromethyl)-1H-benzimidazole
Formula: C₈H₃Cl₂F₃N₂
Storage Temperature: Ambient
MDL Number: MFCD01658509
CAS Number: 2338-25-2
UN: 2811
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR