1-Ethyl-3-(trifluoromethyl)benzene

Supplier: APOLLO SCIENTIFIC
Warning

PC450011-1G PC450011-250MG
77415-448EA 756.98 USD
77415-448 77415-450
1-Ethyl-3-(trifluoromethyl)benzene
1-Ethyl-3-(trifluoromethyl)benzene
Formula: C₉H₉F₃
MW: 174.163 g/mol
Boiling Pt: 52 °C (6 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD24842561
CAS Number: 27190-70-1
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR