1-(Difluoromethyl)-1H-pyrazole

Supplier: APOLLO SCIENTIFIC
Warning

PC411300-1G PC411300-5G
77415-148EA 125.6 USD
77415-148 77415-150
1-(Difluoromethyl)-1H-pyrazole
1-(Difluoromethyl)-1H-pyrazole
Formula: C₄H₄F₂N₂
Storage Temperature: Ambient
MDL Number: MFCD04971168
CAS Number: 908014-77-7
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR