2-Bromo-3-fluorobenzenesulfonyl chloride

Supplier: APOLLO SCIENTIFIC
Danger

PC3705-1G PC3705-250MG
77414-824EA 292.4 USD
77414-824 77414-826
2-Bromo-3-fluorobenzenesulfonyl chloride
2-Bromo-3-fluorobenzenesulfonyl chloride
Formula: C₆H₃BrClFO₂S
Storage Temperature: Ambient
MDL Number: MFCD11101430
CAS Number: 1065076-31-4
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR