2,2,2-Trifluoro-1-(trifluoromethyl)ethylamine 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 1,1,1,3,3,3-Hexafluoropropan-2-amine, 2-Amino-2H-hexafluoropropane

PC3608-1G PC3608-5G
77414-778EA 149.42 USD
77414-778 77414-780 77414-782
2,2,2-Trifluoro-1-(trifluoromethyl)ethylamine 97%
2,2,2-Trifluoro-1-(trifluoromethyl)ethylamine
Formula: C₃H₃F₆N
MW: 167.053 g/mol
Boiling Pt: 57…58 °C
Density: 1.5442 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD08460498
CAS Number: 1619-92-7
UN: 2920
ADR: 8+3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR