3-Fluoro-5-iodobenzyl bromide 95%

Supplier: APOLLO SCIENTIFIC
Danger

PC32739-1G PC32739-250MG PC32739-5G
77414-628EA 210.9 USD
77414-628 77414-630 77414-632
3-Fluoro-5-iodobenzyl bromide 95%
3-Fluoro-5-iodobenzyl bromide
Formula: C₇H₅BrFI
Storage Temperature: Ambient
MDL Number: MFCD18396960
CAS Number: 1261759-75-4
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR