2-(Methylthio)-4-(trifluoromethyl)pyrimidine

Supplier: APOLLO SCIENTIFIC

PC31716-1G PC31716-5G
77414-554EA 176.26 USD
77414-554 77414-556
2-(Methylthio)-4-(trifluoromethyl)pyrimidine
2-(Methylthio)-4-(trifluoromethyl)pyrimidine
Formula: C₆H₅F₃N₂S
MW: 194.178 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00209107
CAS Number: 136547-20-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR