4-Fluoro-3-methylphenylacetonitrile 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: (4-Fluoro-3-methylphenyl)acetonitrile

PC302996-10G PC302996-1G PC302996-5G
77414-306EA 341.3 USD
77414-306 77414-308 77414-310
4-Fluoro-3-methylphenylacetonitrile 98%
4-Fluoro-3-methylphenylacetonitrile
Formula: C₉H₈FN
MW: 149.165 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09832240
CAS Number: 1000548-41-3
UN: 3276
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR