2-Chloro-6-fluoro-3-methoxybenzamide 97%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
PC302445-1G
PC302445-5G
77414-182EA
49.92
USD
77414-182
77414-184
2-Chloro-6-fluoro-3-methoxybenzamide 97%
2-Chloro-6-fluoro-3-methoxybenzamide
Formula:
C₈H₇ClFNO₂ MW: 203.598 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06660325 CAS Number: 886761-58-6 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...