2,6-Difluoro-3-methoxyphenylacetonitrile 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC302162-1G PC302162-5G
77414-122EA 99.84 USD
77414-122 77414-124
2,6-Difluoro-3-methoxyphenylacetonitrile 98%
2,6-Difluoro-3-methoxyphenylacetonitrile
Formula: C₉H₇F₂NO
Storage Temperature: Ambient
MDL Number: MFCD04115909
CAS Number: 886498-69-7
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR