2-Chloro-6-(trifluoromethyl)phenylacetonitrile 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC302025-1G PC302025-25G PC302025-5G
77414-104EA 121.24 USD
77414-104 77414-106 77414-108
2-Chloro-6-(trifluoromethyl)phenylacetonitrile 97%
2-Chloro-6-(trifluoromethyl)phenylacetonitrile
Formula: C₉H₅ClF₃N
MW: 219.591 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06660233
CAS Number: 633317-69-8
UN: 3439
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR