1-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-1H-indole-5-carbonitrile

Supplier: APOLLO SCIENTIFIC

PC300998-250MG PC300998-1G
77414-074EA 135.5 USD
77414-074 77414-072
1-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-1H-indole-5-carbonitrile
1-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-1H-indole-5-carbonitrile
Formula: C₁₅H₇ClF₃N₃
Storage Temperature: Ambient
MDL Number: MFCD31630105
CAS Number: 151387-94-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR