Ethyl 1-(4-bromo-2-fluorophenyl)-5-oxopyrrolidine-3-carboxylate

Supplier: APOLLO SCIENTIFIC

PC300997-1G PC300997-250MG
77414-068EA 320.92 USD
77414-068 77414-070
Ethyl 1-(4-bromo-2-fluorophenyl)-5-oxopyrrolidine-3-carboxylate
Ethyl 1-(4-bromo-2-fluorophenyl)-5-oxopyrrolidine-3-carboxylate
Formula: C₁₃H₁₃BrFNO₃
Storage Temperature: Ambient
MDL Number: MFCD16630891
CAS Number: 1239731-93-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR