Methyl 2,5-difluoro-4-(4-methylpiperazin-1-yl)benzoate

Supplier: APOLLO SCIENTIFIC

PC300788-1G PC300788-250MG PC300788-5G
77414-034EA 275.08 USD
77414-034 77414-036 77414-038
Methyl 2,5-difluoro-4-(4-methylpiperazin-1-yl)benzoate
Methyl 2,5-difluoro-4-(4-methylpiperazin-1-yl)benzoate
Formula: C₁₃H₁₆F₂N₂O₂
MW: 270.275 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28103349
CAS Number: 1858251-84-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR