Methyl 2,5-difluoro-4-morpholin-4-ylbenzoate

Supplier: APOLLO SCIENTIFIC

PC300786-1G PC300786-250MG
77414-026EA 275.08 USD
77414-026 77414-028
Methyl 2,5-difluoro-4-morpholin-4-ylbenzoate
Methyl 2,5-difluoro-4-morpholin-4-ylbenzoate
Formula: C₁₂H₁₃F₂NO₃
MW: 257.233 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28103347
CAS Number: 1858251-27-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR