3-Chloro-2-isothiocyanato-5-(trifluoromethyl)pyridine

Supplier: APOLLO SCIENTIFIC
Danger

PC28287-1G PC28287-250MG
77413-890EA 447.26 USD
77413-890 77413-892
3-Chloro-2-isothiocyanato-5-(trifluoromethyl)pyridine
3-Chloro-2-isothiocyanato-5-(trifluoromethyl)pyridine
Formula: C₇H₂ClF₃N₂S
MW: 238.617 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22557535
CAS Number: 1360887-00-8
UN: 2923
ADR: 8+6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR