1,1,2,2-Tetrafluoro-2-(1,1,2,2-tetrafluoroethoxy)ethanesulfonyl fluoride 95%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 5H-Octafluoro-3-oxapentanesulphonyl fluoride

PC2511-25G PC2511-5G PC2511-1G
77413-674EA 2548.06 USD
77413-674 77413-676 77413-672
1,1,2,2-Tetrafluoro-2-(1,1,2,2-tetrafluoroethoxy)ethanesulfonyl fluoride 95%
1,1,2,2-Tetrafluoro-2-(1,1,2,2-tetrafluoroethoxy)ethanesulfonyl fluoride
Formula: C₄HF₉O₃S
Boiling Pt: 70 °C (400 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD07778617
CAS Number: 104729-49-9
UN: 2920
ADR: 8+3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR