2-(Trifluoromethyl)styrene

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 1-(Trifluoromethyl)-2-vinylbenzene, 2-Vinylbenzotrifluoride, 1-Ethenyl-2-(trifluoromethyl)benzene

PC1998-1G PC1998-5G
77413-438EA 72 USD
77413-438 77413-440
2-(Trifluoromethyl)styrene
2-(Trifluoromethyl)styrene
Formula: C₉H₇F₃
MW: 172.147 g/mol
Boiling Pt: 61 °C
Density: 1.164 g/cm³ (25 °C)
Flash Pt: 42 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00035984
CAS Number: 395-45-9
EINECS: 206-902-2
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR