2-Chloro-2,2-difluoroethane-1,1-diol 95%

Supplier: APOLLO SCIENTIFIC
Warning

PC1728-1G PC1728-5G
77413-290EA 83.54 USD
77413-290 77413-292
2-Chloro-2,2-difluoroethane-1,1-diol 95%
2-Chloro-2,2-difluoroethane-1,1-diol
Formula: C₂H₃ClF₂O₂
MW: 132.494 g/mol
Density: 1.384 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00042464
CAS Number: 63034-47-9
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR