2-Fluoropropan-1-ol 97%

Supplier: APOLLO SCIENTIFIC
Warning

PC1014-1G PC1014-250MG PC1014-5G
77412-788EA 181.08 USD
77412-788 77412-790 77412-792
2-Fluoropropan-1-ol 97%
2-Fluoropropan-1-ol
Formula: C₃H₇FO
Boiling Pt: 115 °C
Storage Temperature: Ambient
MDL Number: MFCD07777178
CAS Number: 3824-87-1
UN: 1987
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR