1,1,1-Trifluorobutan-2-ol 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-Hydroxy-1,1,1-trifluorobutane, 1-(Trifluoromethyl)propan-1-ol

PC0984-1G PC0984-5G
77412-754EA 186.42 USD
77412-754 77412-756
1,1,1-Trifluorobutan-2-ol 97%
1,1,1-Trifluorobutan-2-ol
Formula: C₄H₇F₃O
MW: 128.093 g/mol
Boiling Pt: 90.8…91.8 °C (752 mm Hg)
Density: 1.166 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00462509
CAS Number: 431-36-7
UN: 1987
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR