2-Methyl-4-(trifluoromethoxy)benzonitrile 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-(Trifluoromethoxy)-o-tolunitrile, 2-Cyano-5-(trifluoromethoxy)toluene

PC0418-25G PC0418-5G PC0418-1G
77412-554EA 825.17 USD
77412-554 77412-556 77412-552
2-Methyl-4-(trifluoromethoxy)benzonitrile 98%
2-Methyl-4-(trifluoromethoxy)benzonitrile
Formula: C₉H₆F₃NO
Boiling Pt: 64 °C
Storage Temperature: Ambient
MDL Number: MFCD01631543
CAS Number: 261951-92-2
UN: 3276
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR