1-(4-Bromobenzyl)-5-methyl-1H-1,2,3-triazole-4-carboxylic acid 90%

Supplier: APOLLO SCIENTIFIC
Warning

OR9733-1G OR9733-250MG OR9733-50MG
77412-288EA 192.56 USD
77412-288 77412-290 77412-292
1-(4-Bromobenzyl)-5-methyl-1H-1,2,3-triazole-4-carboxylic acid 90%
1-(4-Bromobenzyl)-5-methyl-1H-1,2,3-triazole-4-carboxylic acid
Formula: C₁₁H₁₀BrN₃O₂
MW: 296.12 g/mol
Melting Pt: 191.6…192.6 °C
Storage Temperature: Ambient
MDL Number: MFCD06659088
CAS Number: 845885-94-1
UN: 1325
ADR: 4.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR