6-Chloro-4-iodopyridin-3-amine

Supplier: APOLLO SCIENTIFIC

OR960084-1G OR960084-250MG OR960084-5G
77412-214EA 26.4 USD
77412-214 77412-216 77412-218
6-Chloro-4-iodopyridin-3-amine
6-Chloro-4-iodopyridin-3-amine
Formula: C₅H₄ClIN₂
Storage Temperature: Refrigerator
MDL Number: MFCD11520341
CAS Number: 351227-42-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR