(3R-Cis)-3,6-dimethyl-1,4-dioxane-2,5-dione 98%

Supplier: APOLLO SCIENTIFIC

Synonyms: (3R,6R)-3,6-Dimethyl-1,4-dioxane-2,5-dione

OR959522-10G OR959522-25G OR959522-5G
77412-202EA 83.54 USD
77412-202 77412-204 77412-206
(3R-Cis)-3,6-dimethyl-1,4-dioxane-2,5-dione 98%
(3R-Cis)-3,6-dimethyl-1,4-dioxane-2,5-dione
Formula: C₆H₈O₄
MW: 144.125 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00082566
CAS Number: 13076-17-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR