Hex-5-yn-1-amine 96%

Supplier: APOLLO SCIENTIFIC
Danger

OR939942-1G OR939942-250MG
77412-078EA 516.54 USD
77412-078 77412-080
Hex-5-yn-1-amine 96%
Hex-5-yn-1-amine
Formula: C₆H₁₁N
Boiling Pt: 144 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD09923614
CAS Number: 15252-45-6
UN: 2733
ADR: 3+8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR