Methyl 4-amino-5-chloro-2-methoxybenzoate 95%

Supplier: APOLLO SCIENTIFIC

OR931029-1G OR931029-250MG OR931029-5G
77411-954EA 140.6 USD
77411-954 77411-956 77411-958
Methyl 4-amino-5-chloro-2-methoxybenzoate 95%
Methyl 4-amino-5-chloro-2-methoxybenzoate
Formula: C₉H₁₀ClNO₃
Melting Pt: 134…139 °C
Storage Temperature: Ambient
MDL Number: MFCD01034555
CAS Number: 20896-27-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR