1-Methyl-5-[(methylamino)methyl]-3-phenyl-1H-pyrazole 95%

Supplier: APOLLO SCIENTIFIC
Danger

OR9136-1G OR9136-250MG
77411-688EA 281.2 USD
77411-688 77411-690
1-Methyl-5-[(methylamino)methyl]-3-phenyl-1H-pyrazole 95%
1-Methyl-5-[(methylamino)methyl]-3-phenyl-1H-pyrazole
Formula: C₁₂H₁₅N₃
MW: 201.268 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08271945
CAS Number: 864068-98-4
UN: 3265
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR