5,6-Dimethyl-2-(pyridin-2-yl)pyrimidin-4-ol

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-(4,5-Dimethyl-6-hydroxypyrimidin-2-yl)pyridine, 4,5-Dimethyl-6-hydroxy-2-(pyridin-2-yl)pyrimidine

OR8472-1G OR8472-5G
77411-396EA 275.08 USD
77411-396 77411-398
5,6-Dimethyl-2-(pyridin-2-yl)pyrimidin-4-ol
5,6-Dimethyl-2-(pyridin-2-yl)pyrimidin-4-ol
Formula: C₁₁H₁₁N₃O
MW: 201.225 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00269287
CAS Number: 204394-52-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR