2-Methyl-5-[(morpholin-4-yl)sulfonyl]-3-furoic acid 95+%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 2-Methyl-5-(4-morpholinosulphonyl)furan-3-carboxylic acid

OR8131-1G OR8131-250MG OR8131-5G
77411-348EA 247.58 USD
77411-348 77411-350 77411-352
2-Methyl-5-[(morpholin-4-yl)sulfonyl]-3-furoic acid 95+%
2-Methyl-5-[(morpholin-4-yl)sulfonyl]-3-furoic acid
Formula: C₁₀H₁₃NO₆S
MW: 275.278 g/mol
Melting Pt: 184 °C
Storage Temperature: Ambient
MDL Number: MFCD02180756
CAS Number: 306936-37-8
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR