2-(5-Chloro-1,2,4-thiadiazol-3-yl)thiophene-5-sulfonyl chloride 95%

Supplier: APOLLO SCIENTIFIC
Danger

OR8065-1G OR8065-500MG
77411-340EA 434.02 USD
77411-340 77411-342
2-(5-Chloro-1,2,4-thiadiazol-3-yl)thiophene-5-sulfonyl chloride 95%
2-(5-Chloro-1,2,4-thiadiazol-3-yl)thiophene-5-sulfonyl chloride
Formula: C₆H₂Cl₂N₂O₂S₃
Melting Pt: 71…74 °C
Storage Temperature: Ambient
MDL Number: MFCD02180888
CAS Number: 306937-21-3
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR