2-Chloro-4-(methylsulfonyl)benzaldehyde hydrate 95+%

Supplier: APOLLO SCIENTIFIC
Warning

OR7624-1G OR7624-5G
77411-264EA 251.64 USD
77411-264 77411-266
2-Chloro-4-(methylsulfonyl)benzaldehyde hydrate 95+%
2-Chloro-4-(methylsulfonyl)benzaldehyde hydrate
Formula: C₈H₇ClO₃S·H₂O
MW: 236.673 g/mol
Melting Pt: 111…112 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD08166750
CAS Number: 1171826-50-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR