3-(Phenylamino)-2-(3,4,5-trimethoxybenzyl)acrylonitrile

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 3-Anilino-2-(3,4,5-trimethoxybenzyl)acrylonitrile, Trimethoprim Impurity I

OR7250T-100MG OR7250T-1G OR7250T-500MG
77411-194EA 117.16 USD
77411-194 77411-196 77411-198
3-(Phenylamino)-2-(3,4,5-trimethoxybenzyl)acrylonitrile
3-(Phenylamino)-2-(3,4,5-trimethoxybenzyl)acrylonitrile

A reference standard for the drug Trimethoprim.

Formula: C₁₉H₂₀N₂O₃
Melting Pt: 130…136 °C
Storage Temperature: Ambient
MDL Number: MFCD00269911
CAS Number: 30078-48-9
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR