Thieno[2,3-b]pyrazine-6-carboxylic acid 95%

Supplier: APOLLO SCIENTIFIC
Warning

OR6789-1G OR6789-250MG
77410-902EA 241.46 USD
77410-902 77410-904
Thieno[2,3-b]pyrazine-6-carboxylic acid 95%
Thieno[2,3-b]pyrazine-6-carboxylic acid
Formula: C₇H₄N₂O₂S
MW: 180.184 g/mol
Melting Pt: 220 °C
Storage Temperature: Ambient
MDL Number: MFCD07371382
CAS Number: 59944-79-5
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR