2-Acetamido-1-amino-1,2-dideoxy-β-D-glucopyranose 95%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-Acetamido-2-deoxy-β-D-glucosamine

OR5825T-1G OR5825T-250MG
77410-542EA 330.1 USD
77410-542 77410-544
2-Acetamido-1-amino-1,2-dideoxy-β-D-glucopyranose 95%
2-Acetamido-1-amino-1,2-dideoxy-β-D-glucopyranose

Used in the synthesis of various 1-N-acyl derivatives which model the N-glycosylamine linkage of glycoproteins.

Formula: C₈H₁₆N₂O₅
MW: 220.223 g/mol
Melting Pt: 103…112 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00057751
CAS Number: 4229-38-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR