Tris(N,N-dimethylaminopropyl)amine

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: N1,N1-Bis(3-(Dimethylamino)propyl)-N3,N3-dimethylpropane-1,3-diamine

OR55744-1G OR55744-250MG
77410-522EA 141.62 USD
77410-522 77410-524
Tris(N,N-dimethylaminopropyl)amine
Tris(N,N-dimethylaminopropyl)amine
Formula: C₁₅H₃₆N₄
Storage Temperature: Refrigerator
MDL Number: MFCD00190091
CAS Number: 33329-35-0
UN: 2735
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR