Methyl pent-4-ynoate

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 4-Pentynoic acid methyl ester

OR55646-1G OR55646-25G OR55646-5G
77410-488EA 21.26 USD
77410-488 77410-490 77410-492
Methyl pent-4-ynoate
Methyl pent-4-ynoate
Formula: C₆H₈O₂
MW: 112.127 g/mol
Boiling Pt: 141…142 °C
Storage Temperature: Refrigerator
MDL Number: MFCD12025043
CAS Number: 21565-82-2
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR