3,7-Dioxabicyclo[4.1.0]heptane 95%

Supplier: APOLLO SCIENTIFIC
Warning

OR54673-1G OR54673-250MG
77410-422EA 104.06 USD
77410-422 77410-424
3,7-Dioxabicyclo[4.1.0]heptane 95%
3,7-Dioxabicyclo[4.1.0]heptane
Formula: C₅H₈O₂
Storage Temperature: Refrigerator
MDL Number: MFCD11226853
CAS Number: 286-22-6
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR