2-(Cyclopropyl)benzyl bromide

Supplier: APOLLO SCIENTIFIC
Danger

OR54649-250MG OR54649-1G
77410-414EA 206.82 USD
77410-414 77410-412
2-(Cyclopropyl)benzyl bromide
2-(Cyclopropyl)benzyl bromide
Formula: C₁₀H₁₁Br
MW: 211.098 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD15527184
CAS Number: 1260801-90-8
UN: 1760
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR