3-(Cyclopropyl)benzyl bromide

Supplier: APOLLO SCIENTIFIC
Danger

OR54648-1G OR54648-250MG
77410-408EA 496.16 USD
77410-408 77410-410
3-(Cyclopropyl)benzyl bromide
3-(Cyclopropyl)benzyl bromide
Formula: C₁₀H₁₁Br
Storage Temperature: Refrigerator
MDL Number: MFCD15527185
CAS Number: 1260850-05-2
UN: 1760
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR