3,4-Dihydro-3-oxo-2H-1,4-benzoxazine-6-carbonitrile 95+%

Supplier: APOLLO SCIENTIFIC

Synonyms: 3-Oxo-3,4-dihydro-2H-benzo[b][1,4]oxazine-6-carbonitrile, 3-Oxo-3,4-dihydro-2H-1,4-benzoxazine-6-carbonitrile

OR52436-1G OR52436-250MG
77410-248EA 186.44 USD
77410-248 77410-250
3,4-Dihydro-3-oxo-2H-1,4-benzoxazine-6-carbonitrile 95+%
3,4-Dihydro-3-oxo-2H-1,4-benzoxazine-6-carbonitrile
Formula: C₉H₆N₂O₂
MW: 174.156 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20039908
CAS Number: 134997-74-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR