4-(Tributylstannyl)toluene

Supplier: APOLLO SCIENTIFIC
Danger

OR52031-1G OR52031-250MG
77410-188EA 545.06 USD
77410-188 77410-190
4-(Tributylstannyl)toluene
4-(Tributylstannyl)toluene
Formula: C₁₉H₃₄Sn
Storage Temperature: Refrigerator
MDL Number: MFCD01114692
CAS Number: 31614-66-1
UN: 2788
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR