1-(4-Methoxyphenyl)-cyclopentanemethanamine 97%

Supplier: APOLLO SCIENTIFIC

OR452215-100MG OR452215-250MG
77409-850EA 63.8 USD
77409-850 77409-852
1-(4-Methoxyphenyl)-cyclopentanemethanamine 97%
1-(4-Methoxyphenyl)-cyclopentanemethanamine
Formula: C₁₃H₁₉NO
Storage Temperature: Refrigerator
MDL Number: MFCD01817110
CAS Number: 23528-54-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR