4-Bromo-2-(methylsulfonyl)benzonitrile

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 5-Bromo-2-cyanophenyl methyl sulphone

OR42106-1G OR42106-250MG OR42106-5G
77409-636EA 166.06 USD
77409-636 77409-638 77409-640
4-Bromo-2-(methylsulfonyl)benzonitrile
4-Bromo-2-(methylsulfonyl)benzonitrile
Formula: C₈H₆BrNO₂S
MW: 260.108 g/mol
Melting Pt: 142…144 °C
Storage Temperature: Ambient
MDL Number: MFCD12828670
CAS Number: 1208402-11-2
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR