9-(Bromomethyl)anthracene 95%

Supplier: APOLLO SCIENTIFIC
Danger

OR40727-1G OR40727-250MG OR40727-5G
77409-582EA 144.68 USD
77409-582 77409-584 77409-586
9-(Bromomethyl)anthracene 95%
9-(Bromomethyl)anthracene
Formula: C₁₅H₁₁Br
Storage Temperature: Ambient
MDL Number: MFCD00096212
CAS Number: 2417-77-8
UN: 3077
ADR: 9+EHS,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR