Methyl pent-2-enoate 97%

Supplier: APOLLO SCIENTIFIC
Danger

OR40582-25G OR40582-5G
77409-556EA 79.38 USD
77409-556 77409-558
Methyl pent-2-enoate 97%
Methyl pent-2-enoate
Formula: C₆H₁₀O₂
Boiling Pt: 81…82 °C (45 mm Hg)
Storage Temperature: Freezer
MDL Number: MFCD00137611
CAS Number: 818-59-7
UN: 1993
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR